
5-Bromo-2,3-dihydrobenzofuran-7-carboxylic acid ≥97.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:41177-72-4
:
Formula:C₉H₇BrO₃
MDL Number:MFCD00191391
:243.06 g/mol
:GHS07
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (GC):min. 96.0 %
- Purity (Neutralization titration):min. 97.0 %
- Melting point:230.0 to 235.0 deg-C
Specification Test Results
Frequently Bought Together