
1,2-Bis(2-iodoethoxy)ethane ≥96.0% (by GC) stabilised
Catalog # TCIAB1687-10G
Supplier: TCI
CAS Number:
Specifications
- Pk:10 g
- Seq:3236
Specifications
Specification Test Results
- Appearance:Light yellow to Brown clear liquid
- Purity (GC):min. 96.0 %
Specification Test Results
About this item
:
:369.97 g/mol
:Stabilised with copper chip
:GHS07
CAS Number:36839-55-1
Formula:C₆H₁₂I₂O₂
MDL Number:MFCD00075008