Order Entry
Italy
ContactUsLinkComponent
N-Benzylquinidinium chloride ≥98.0% (by titrimetric analysis) chiral phase-transfer catalyst
N-Benzylquinidinium chloride ≥98.0% (by titrimetric analysis) chiral phase-transfer catalyst
Catalog # TCIAB1684-5G
Supplier:  TCI
CAS Number:  
N-Benzylquinidinium chloride ≥98.0% (by titrimetric analysis) chiral phase-transfer catalyst
Catalog # TCIAB1684-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    White to Orange to Green powder to crystal
  • Purity (Nonaqueous Titration):
    min. 98.0 %
  • Specific rotation [a]20/D:
    +200 to +230 deg (C=0.64, H2O)

Specification Test Results

About this item

 :
 :451.01 g/mol
CAS Number:77481-82-4
Formula:C₂₇H₃₁ClN₂O₂
MDL Number:MFCD00198106