Order Entry
Italy
ContactUsLinkComponent
N-(tert-Butoxycarbonyl)-O-benzyl-L-threonine ≥98.0% (by HPLC, titration analysis)
N-(tert-Butoxycarbonyl)-O-benzyl-L-threonine ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAB1630-5G
Supplier:  TCI
CAS Number:  
N-(tert-Butoxycarbonyl)-O-benzyl-L-threonine ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAB1630-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Neutralization titration):
    min. 98.0 %
  • Melting point:
    114.0 to 118.0 deg-C
  • Specific rotation [a]20/D:
    +16.0 to +17.0 deg (C=1, MeOH)

Specification Test Results

About this item

 :
 :309.36 g/mol
EINECS:239-304-5
CAS Number:15260-10-3
Formula:C₁₆H₂₃NO₅
MDL Number:MFCD00066062