

1,1'-Bis(2,4-dinitrophenyl)-4,4'-bipyridinium dichloride ≥97.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:41168-79-0
:
Formula:C₂₂H₁₄Cl₂N₆O₈
MDL Number:MFCD00142352
:561.29 g/mol
UN:1325
ADR:4.1,III
:GHS02
:GHS08
Specification Test Results
- Appearance:Light yellow to Yellow to Orange powder to crystal
- Purity (HPLC):min. 97.0 area%
Specification Test Results
Frequently Bought Together