
2,2-Bis(4-isocyanatophenyl)hexafluoropropane ≥98.0% (by GC)
Catalog # TCIAB1509-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Purity (GC):min. 98.0 %
Specification Test Results
About this item
:
:386.25 g/mol
Storage Temperature:Refrigerator
UN:2206
ADR:6.1,III
:GHS06
CAS Number:10224-18-7
Formula:C₁₇H₈F₆N₂O₂
MDL Number:MFCD00142759