
2,6-Bis(chloromethyl)pyridine ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:3099-28-3
:
Formula:C₇H₇Cl₂N
MDL Number:MFCD00142844
:176.04 g/mol
Boiling Pt:110°C
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:73.0 to 77.0 deg-C
- Solubility in Methanol:very faint turbidity
Specification Test Results
Frequently Bought Together