2,2'-Bi(1,8-naphthyridine)
Catalog # TCIAB1423-100MG
Supplier: TCI
CAS Number:
Specifications
- Pk:100 mg
- Seq:2440
Specifications
Specification Test Results
- Appearance:Yellow to Brown to Dark green powder to crystal
Specification Test Results
About this item
:
:258.28 g/mol
Melting Pt:227°C
CAS Number:69110-33-4
Formula:C₁₆H₁₀N₄
MDL Number:MFCD00142731