2,2-Bis(4-methylphenyl)hexafluoropropane ≥98.0% (by GC)
Catalog # TCIAB1417-10G
Supplier: TCI
CAS Number:
Specifications
- Pk:10 g
- Seq:3236
Specifications
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:82.0 to 85.0 deg-C
- Solubility in Acetone:almost transparency
Specification Test Results
About this item
:
:332.29 g/mol
Boiling Pt:112°C
CAS Number:1095-77-8
Formula:C₁₇H₁₄F₆
MDL Number:MFCD00042597