
2,2-Bis[4-(4-aminophenoxy)phenyl]hexafluoropropane ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:69563-88-8
:
Formula:C₂₇H₂₀F₆N₂O₂
MDL Number:MFCD00015723
:518.46 g/mol
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:162.0 to 165.0 deg-C
- Solubility in Methanol:almost transparency
Specification Test Results
Frequently Bought Together