N,N'-Bis(4-chlorophenyl)-N,N'-diphenyl-1,4-phenylenediamine ≥98.0% (by GC)
Catalog # TCIAB1336-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Elemental analysis (Nitrogen):5.50 to 6.00 %
- Melting point:204.0 to 208.0 deg-C
Specification Test Results
About this item
:
:481.42 g/mol
CAS Number:113703-66-5
Formula:C₃₀H₂₂Cl₂N₂
MDL Number:MFCD00060108