Order Entry
Italy
ContactUsLinkComponent
N-(tert-Butoxycarbonyl)-L-phenylalanine ≥99.0% (by titrimetric analysis)
N-(tert-Butoxycarbonyl)-L-phenylalanine ≥99.0% (by titrimetric analysis)
Supplier:  TCI
Certificates
N-(tert-Butoxycarbonyl)-L-phenylalanine ≥99.0% (by titrimetric analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (Neutralization titration):
    min. 99.0 %
  • Melting point:
    86.0 to 91.0 deg-C
  • Specific rotation [a]20/D:
    +23.0 to +26.0 deg (C=1, EtOH)
  • Solubility in Methanol:
    almost transparency

Specification Test Results

Related Information

Frequently Bought Together