
[5,5'-Biisobenzofuran]-1,1',3,3'-tetraone ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:2420-87-3
:
Formula:C₁₆H₆O₆
MDL Number:MFCD00039140
:294.2200 g/mol
Melting Pt:303°C
EINECS:219-342-9
:GHS07
Specification Test Results
- Appearance:White to Green to Brown powder to crystal
- Purity (Morpholine Method):min. 98.0 %
- Solubility in hot DMF:almost transparency
Specification Test Results
Frequently Bought Together