Order Entry
Italy
ContactUsLinkComponent
Bis(trimethylsilyl)malonate ≥97.0% (by GC)
Flammable
Bis(trimethylsilyl)malonate ≥97.0% (by GC)
Catalog # TCIAB1302-5ML
Supplier:  TCI
CAS Number:  
Flammable
Bis(trimethylsilyl)malonate ≥97.0% (by GC)
Catalog # TCIAB1302-5ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    5 mL
  • Seq:
    2210

Specifications

Specification Test Results

  • Appearance:
    Colorless to Light yellow clear liquid
  • Purity (GC):
    min. 97.0 %

Specification Test Results

About this item

 :
Boiling Pt:102°C
Flash Pt:24°C
Storage Temperature:Refrigerator
UN:3272
ADR:3,III
 :248.43 g/mol
EINECS:242-342-5
 :GHS02
CAS Number:18457-04-0
Formula:C₉H₂₀O₄Si₂
MDL Number:MFCD00009631