Order Entry
Italy
ContactUsLinkComponent
 
1,8-Bis(2-pyridyl)-3,6-dithiaoctane ≥98.0% (by GC, titration analysis)
 :  TCI
 
1,8-Bis(2-pyridyl)-3,6-dithiaoctane ≥98.0% (by GC, titration analysis)
 :  TCI

 

  • Appearance:
    White to Light yellow to Dark green powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Purity (Nonaqueous Titration):
    min. 98.0 %
  • Melting point:
    48.0 to 51.0 deg-C

 

Frequently Bought Together