2,2'-Bis(2-chlorophenyl)-4,4',5,5'-tetraphenyl-1,2'-biimidazole ≥97.0% (by titrimetric analysis) photopolymerization initiator
Catalog # TCIAB1225-25G
Supplier: TCI
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:Light yellow to Yellow to Green powder to crystal
- Purity (Nonaqueous Titration):min. 97.0 %
- Melting point:196.0 to 201.0 deg-C
Specification Test Results
About this item
:
:659.61 g/mol
EINECS:230-555-6
CAS Number:7189-82-4
:MFCD09028023
Formula:C₄₂H₂₈Cl₂N₄
MDL Number:MFCD00060081