Bis[(2R,3aS,4R,7aS)octahydro-7,8,8-trimethyl-4,7-methanobenzofuran-2-yl]ether ≥98.0% (by HPLC) for optical resolution
Catalog # TCIAB1219-100MG
Supplier: TCI
CAS Number:
Specifications
- Pk:100 mg
- Seq:2440
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Melting point:147.0 to 151.0 deg-C
- Specific rotation [a]20/D:+200.0 to +205.0 deg (C=2, THF)
Specification Test Results
About this item
:
:374.57 g/mol
CAS Number:87248-50-8
Formula:C₂₄H₃₈O₃
MDL Number:MFCD00074783