
4,4'-Biphenyldicarbonitrile ≥97.0% (by GC)
Catalog # TCIAB1181-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Amber powder to crystal
- Purity (GC):min. 97.0 %
- Melting point:235.0 to 240.0 deg-C
Specification Test Results
About this item
:
:204.23 g/mol
EINECS:216-468-6
:GHS07
CAS Number:1591-30-6
Formula:C₁₄H₈N₂
MDL Number:MFCD00013805