Order Entry
Italy
ContactUsLinkComponent
 
 
4,4'-Biphenyldisulphonyl Chloride ≥97.0% (by titrimetric analysis)
 :  TCI
 
 
4,4'-Biphenyldisulphonyl Chloride ≥97.0% (by titrimetric analysis)
 :  TCI

 

  • Appearance:
    White to Orange to Green powder to crystal
  • Purity (Argentmetric Titration):
    min. 97.0 %
  • Melting point:
    207.0 to 212.0 deg-C

 

 

Frequently Bought Together