3,9-Bis(1,1-dimethyl-2-hydroxyethyl)-2,4,8,10-tetraoxaspiro[5.5]undecane ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:1455-42-1
:
Formula:C₁₅H₂₈O₆
MDL Number:MFCD00059794
:304.38 g/mol
EINECS:485-230-3
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:196.0 to 202.0 deg-C
- Solubility in hot Methanol:almost transparency
Specification Test Results
Frequently Bought Together