1-Benzyl-1,4-dihydronicotinamide ≥95.0% (by HPLC, total nitrogen)
Supplier: TCI
Certificates
About this item
CAS Number:952-92-1
:
Formula:C₁₃H₁₄N₂O
MDL Number:MFCD00059769
:214.27 g/mol
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:Light yellow to Brown powder to crystal
- Purity (HPLC):min. 95.0 area%
- Purity (with Total Nitrogen):min. 95.0 %
- Melting point:118.0 to 123.0 deg-C
- Solubility in Methanol:almost transparency
Specification Test Results
Frequently Bought Together