



tert-Butylmagnesium chloride 23% ∼2 mol/L in THF
Catalog # TCIAB1148-250G
Supplier: TCI
CAS Number:
Specifications
- Pk:250 g
- Seq:3545
Specifications
Specification Test Results
- Concentration (sec Butanol method):23.0 to 26.0 w/w%
Specification Test Results
About this item
:
:116.87 g/mol
EINECS:211-638-6
UN:3399
ADR:4.3,I
:GHS02
:GHS05
:GHS07
:GHS08
CAS Number:677-22-5
Formula:C₄H₉ClMg
MDL Number:MFCD00000465