
2,6-Bis(4-azidobenzylidene)cyclohexanone ≥85.0% (by HPLC) wetted with ca. 30% water (unit weight on dry weight basis) research for photosensitive material
Supplier: TCI
Certificates
About this item
CAS Number:20237-98-3
:
Formula:C₂₀H₁₆N₆O
MDL Number:MFCD00059878
:356.39 g/mol
EINECS:243-625-6
UN:1325
ADR:4.1,III
:GHS02
Specification Test Results
- Appearance:Light yellow to Brown powder to crystal
- Purity (HPLC):min. 85.0 area%
Specification Test Results
Frequently Bought Together