

1,4-Bis(aminomethyl)cyclohexane (cis and trans mixture) cis- and trans-mixture ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:2549-93-1
:
Formula:C₈H₁₈N₂
MDL Number:MFCD00039463
:142.25 g/mol
Flash Pt:106°C
EINECS:219-840-6
UN:2922
ADR:8,II
:GHS05
:GHS06
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 % (total of isomers)
- Purity (Neutralization titration):min. 98.0 %
Specification Test Results
Frequently Bought Together