2,3-Bis(4-ethylphenyl)-5-phenyltetrazolium chloride ≥98.0% (by HPLC)
Catalog # TCIAB1053-100MG
Supplier: TCI
CAS Number:
Specifications
- Pk:100 mg
- Seq:2440
Specifications
Specification Test Results
- Appearance:White to Light yellow to Light red powder to crystal
- Purity (HPLC):min. 98.0 area%
Specification Test Results
About this item
:
:390.92 g/mol
CAS Number:104497-78-1
Formula:C₂₃H₂₃ClN₄
MDL Number:MFCD00060006