Order Entry
Italy
ContactUsLinkComponent
1-Bromo-2-butanol ≥70% (by GC) (contains ca. 30% 2-Bromo-1-butanol)
Flammable
1-Bromo-2-butanol ≥70% (by GC) (contains ca. 30% 2-Bromo-1-butanol)
Catalog # TCIAB0932-25G
Supplier:  TCI
CAS Number:  
Flammable
1-Bromo-2-butanol ≥70% (by GC) (contains ca. 30% 2-Bromo-1-butanol)
Catalog # TCIAB0932-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    Colorless to Light yellow to Light orange clear liquid
  • Purity (GC):
    min. 70.0 %

Specification Test Results

About this item

 :
 :153.02 g/mol
UN:1993
ADR:3,III
 :GHS02
CAS Number:2482-57-7
Formula:C₄H₉BrO
MDL Number:MFCD00059883