

N,O-Bis(trimethylsilyl)trifluoroacetamide kit
Catalog # TCIAB0912-1KIT
Supplier: TCI
CAS Number:
Specifications
- Pk:1 KIT
- Seq:0075
- Pk Info:1 mL × 8
Specifications
Specification Test Results
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (GC):min. 95.0 %
Specification Test Results
About this item
:
:257.4 g/mol
Boiling Pt:142°C
Flash Pt:24°C
Storage Temperature:Refrigerator
EINECS:247-103-9
UN:2924
ADR:3,III
:GHS02
:GHS05
CAS Number:25561-30-2
Formula:C₈H₁₈F₃NOSi₂
MDL Number:MFCD00008269