Order Entry
Italy
ContactUsLinkComponent
Boron trifluoridedimethanol complex
Flammable
Corrosive
Irritant
Health
Boron trifluoridedimethanol complex
Supplier:  TCI
Certificates
Flammable
Corrosive
Irritant
Health
Boron trifluoridedimethanol complex
Supplier:  TCI
Certificates

Specification Test Results

  • Appearance:
    Colorless to Light orange to Yellow clear liquid
  • Concentration (Gravimetric Method):
    49.0 to 54.0 w/w%

Specification Test Results

Related Information

Frequently Bought Together