Order Entry
Italy
ContactUsLinkComponent
2,2'-Bi-4-lepidine ≥98.0% (by titrimetric analysis)
Irritant
2,2'-Bi-4-lepidine ≥98.0% (by titrimetric analysis)
Catalog # TCIAB0841-100MG
Supplier:  TCI
CAS Number:  
Irritant
2,2'-Bi-4-lepidine ≥98.0% (by titrimetric analysis)
Catalog # TCIAB0841-100MG
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Seq:
    2440

Specifications

Specification Test Results

  • Appearance:
    White to Light yellow to Green powder to crystal
  • Purity (HPLC):
    min. 97.0 area%
  • Purity (Nonaqueous Titration):
    min. 98.0 %

Specification Test Results

About this item

 :
 :284.36 g/mol
 :GHS07
CAS Number:7654-51-5
Formula:C₂₀H₁₆N₂
MDL Number:MFCD00059753