Order Entry
Italy
ContactUsLinkComponent
 
Diethyl-4,4'-biphenyldicarboxylate ≥98.0% (by GC, titration analysis)
 :  TCI
 
Diethyl-4,4'-biphenyldicarboxylate ≥98.0% (by GC, titration analysis)
 :  TCI

 

  • Appearance:
    White to Light yellow powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Purity (Ester Value):
    min. 98.0 %
  • Melting point:
    112.0 to 116.0 deg-C
  • Solubility in hot Toluene:
    almost transparency

 

Frequently Bought Together