Acid orange 17
25 g
Catalog # TCIAB0778-25G
Supplier: TCI
Jump to:
Product Details & Documents
Synonyms: Brilliant Orange H
- CAS Number:52749-23-2
- Formula:C₁₈H₁₆N₂O₄S
- MDL Number:MFCD00059907
- Molecular Weight:378.38 g/mol
- EINECS:258-153-6
Specifications
Specification Test Results
- Appearance:Light yellow to Yellow to Orange powder to crystal
- Lambda max.:490.0 to 494.0 nm (H2O)
- Absorbance (E1%1cm):min. 450 (H2O, 490.0 to 494.0 nm)
Specifications
- Pk:25 g



