

Brucine hydrate ≥99.0% (by titrimetric analysis)
Catalog # TCIAB0670-25G
Supplier: TCI
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 99.0 % (calcd.on anh.substance)
- Specific rotation [a]20/D:-75.0 to -80.0 deg (C=1, EtOH)
- Solubility in EtOH:almost transparency
Specification Test Results
About this item
:
:394.47 g/mol
Merck Index:14,01455
UN:1570
ADR:6.1,I
:GHS06
:GHS08
:5892-11-5
Formula:C₂₃H₂₆N₂O₄