Order Entry
Italy
ContactUsLinkComponent
1-Bromo-2-propanol ≥75.0% (by GC) (contains ca. 20% 2-Bromo-1-propanol) stabilised
Flammable
Corrosive
1-Bromo-2-propanol ≥75.0% (by GC) (contains ca. 20% 2-Bromo-1-propanol) stabilised
Catalog # TCIAB0640-25G
Supplier:  TCI
CAS Number:  
Flammable
Corrosive
1-Bromo-2-propanol ≥75.0% (by GC) (contains ca. 20% 2-Bromo-1-propanol) stabilised
Catalog # TCIAB0640-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    Colorless to Light yellow clear liquid
  • Purity (GC):
    min. 75.0 %

Specification Test Results

About this item

 :
 :138.99 g/mol
Flash Pt:54°C
Storage Temperature:Refrigerator
UN:2920
ADR:8,II
 :Stabiised with magnesium oxide (MgO)
EINECS:243-225-1
 :GHS02
 :GHS05
CAS Number:19686-73-8
Formula:BrH₂CCH(OH)CH₃
MDL Number:MFCD00004529