Bis(2-hydroxyethyl)dithiocarbamic acid zinc(II) salt ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:19163-92-9
:
Formula:C₅H₁₁NO₂S₂
MDL Number:MFCD00070508
:425.90 g/mol
EINECS:242-852-8
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Chelometric Titration):min. 98.0 %
Specification Test Results
Frequently Bought Together