MTT formazan ≥98.0% (by HPLC)
TCIAB0275-100MG
: TCI
:
- Pk:100 mg
- Seq:2440
- Appearance:Black powder to crystal
- Purity (HPLC):min. 98.0 area%
- Melting point:161.0 to 169.0 deg-C
:
:335.43 g/mol
:245-566-1
:23305-68-2
:C₁₈H₁₇N₅S
:MFCD00080731