

(±)-2-Bromopentane ≥85.0% (by GC) (contains ca. 12% 3-Bromopentane)
Supplier: TCI
Certificates
About this item
CAS Number:107-81-3
:
Formula:H₃CBrCH(CH₂)₂CH₃
MDL Number:MFCD00000160
:151.05 g/mol
Boiling Pt:117°C
Flash Pt:32°C
EINECS:203-521-3
UN:2343
ADR:3,II
:GHS02
:GHS07
Specification Test Results
- Appearance:Colorless to Light orange to Yellow clear liquid
- Purity (GC):min. 85.0 %
Specification Test Results
Frequently Bought Together