Order Entry
Italy
ContactUsLinkComponent
Trimethyl borate 63 - 65% in methanol
Flammable
Irritant
Health
Trimethyl borate 63 - 65% in methanol
Supplier:  TCI
Certificates
undefined
Flammable
Irritant
Health
Trimethyl borate 63 - 65% in methanol
Supplier:  TCI

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Concentration (Neutralization titration):
    63.0 to 65.0 w/w%

Specification Test Results

Related Information

Frequently Bought Together