1,2-Benzanthraquinone ≥95.0% (by HPLC)
Catalog # TCIAB0018-5G
Supplier: TCI
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:Light yellow to Amber to Dark green powder to crystal
- Purity (HPLC):min. 95.0 area%
- Melting point:166.0 to 171.0 deg-C
- Solubility in Chloroform:very faint turbidity
Specification Test Results
About this item
:
:258.28 g/mol
EINECS:219-693-8
CAS Number:2498-66-0
Formula:C₁₈H₁₀O₂
MDL Number:MFCD00003596