Benzaldehyde-4-nitrophenylhydrazone ≥98.0% (by HPLC)
TCIAB0009-25G
: TCI
:
- Pk:25 g
- Seq:3308
- Appearance:Light yellow to Brown powder to crystal
- Purity (HPLC):min. 98.0 area%
- Melting point:193.0 to 197.0 deg-C
- Solubility in Methanol:almost transparency
:
:241.25 g/mol
:3078-09-9
:C₁₃H₁₁N₃O₂
:MFCD00024596