Dansylamino-PITC ≥98.0% (by HPLC, total nitrogen) fluorescent coupling reagent for Edman degradation
Catalog # TCIAA5354-100MG
Supplier: TCI
CAS Number:
Specifications
- Pk:100 mg
- Seq:2440
Specifications
Specification Test Results
- Appearance:Light yellow to Amber to Dark green powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 98.0 %
- Melting point:127.0 to 131.0 deg-C
Specification Test Results
About this item
:
:383.48 g/mol
Storage Temperature:Refrigerator
CAS Number:102417-94-7
Formula:C₁₉H₁₇N₃O₂S₂
MDL Number:MFCD00077739