H-Resorcinol ≥97.0% (by HPLC) spectrophotometric reagent for the determination of B by FIA
Catalog # TCIAA5017-1G
Supplier: TCI
CAS Number:
Specification Test Results
- Appearance:Red to Dark blue to Black powder to crystal
- Purity (HPLC):min. 97.0 area%
- Suitability for boron-analysis:to pass test