
2,2'-Dihydroxyazobenzene ≥98.0% (by HPLC, titration analysis) for spectrophotometry, for fluorimetry
Supplier: TCI
Certificates
About this item
CAS Number:2050-14-8
:
Formula:C₁₂H₁₀N₂O₂
MDL Number:MFCD00002182
:214.22 g/mol
EINECS:218-082-3
:GHS07
Specification Test Results
- Appearance:Light yellow to Amber to Dark green powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 %
- Melting point:170.0 to 179.0 deg-C
Specification Test Results
Frequently Bought Together