
2-Acetyl-3,5-dimethylpyrazine ≥98.0% (by GC) (contains 2-Acetyl-3,6-dimethylpyrazine)
Supplier: TCI
Certificates
About this item
CAS Number:54300-08-2
:
Formula:C₈H₁₀N₂O
MDL Number:MFCD00055023
:150.18 g/mol
Flash Pt:91°C
EINECS:259-076-0
:GHS07
Specification Test Results
- Appearance:Colorless to Light orange to Yellow clear liquid
- Purity (GC):min. 98.0 % (total of isomer)
- Specific gravity (20/20):1.0740 to 1.0840
Specification Test Results
Frequently Bought Together