
Adamantane-1,3-diamine ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:10303-95-4
:
Formula:C₁₀H₁₈N₂
MDL Number:MFCD01831403
:166.27 g/mol
UN:3259
ADR:8,II
:GHS05
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
Specification Test Results
Frequently Bought Together