Order Entry
Italy
ContactUsLinkComponent
Amantadine ≥98.0% (by GC, titration analysis), purified by sublimation
Corrosive
Irritant
Health
Amantadine ≥98.0% (by GC, titration analysis), purified by sublimation
Catalog # TCIAA3234-1G
Supplier:  TCI
CAS Number:  
Corrosive
Irritant
Health
Amantadine ≥98.0% (by GC, titration analysis), purified by sublimation
Catalog # TCIAA3234-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Purity (Nonaqueous Titration):
    min. 98.0 %

Specification Test Results

About this item

 :
 :151.25 g/mol
Merck Index:14,00374
EINECS:212-201-2
UN:3259
ADR:8,II
 :GHS05
 :GHS07
 :GHS08
CAS Number:768-94-5
Formula:C₁₀H₁₇N
MDL Number:MFCD00074732