
N-Acetyl-di-O-acetylganciclovir ≥98.0% (by HPLC, total nitrogen)
Catalog # TCIAA3228-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 98.0 %
- Melting point:172.0 to 176.0 deg-C
Specification Test Results
About this item
:
:381.35 g/mol
:GHS08
CAS Number:86357-14-4
Formula:C₁₅H₁₉N₅O₇
MDL Number:MFCD08457700