
Acridine-3,6-diamine ≥96.0% (by HPLC, titration analysis)
: TCI
:92-62-6
:
:C₁₃H₁₁N₃
:MFCD00005030
:209.25 g/mol
:14,07771
:202-172-4
:GHS07
- Appearance:Light yellow to Brown powder to crystal
- Purity (HPLC):min. 96.0 area%
- Purity (Nonaqueous Titration):min. 96.0 %
Frequently Bought Together