Order Entry
Italy
ContactUsLinkComponent
Adrafinil ≥98.0% (by total nitrogen basis)
Irritant
Adrafinil ≥98.0% (by total nitrogen basis)
Supplier:  TCI
Certificates
Irritant
Adrafinil ≥98.0% (by total nitrogen basis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Light yellow to Light orange powder to crystal
  • Purity (HPLC):
    min. 96.0 area%
  • Purity (with Total Nitrogen):
    min. 98.0 %
  • Melting point:
    151.0 to 154.0 deg-C

Specification Test Results

Frequently Bought Together