Order Entry
Italy
ContactUsLinkComponent
2-Amino-5-methoxypyridine ≥97.0% (by GC, titration analysis)
Irritant
2-Amino-5-methoxypyridine ≥97.0% (by GC, titration analysis)
Catalog # TCIAA3168-1G
Supplier:  TCI
CAS Number:  
Irritant
2-Amino-5-methoxypyridine ≥97.0% (by GC, titration analysis)
Catalog # TCIAA3168-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    1 g/mol
  • Seq:
    9220

Specifications

Specification Test Results

  • Appearance:
    White or Colorless to Light orange to Yellow powder to lump to clear liquid
  • Purity (GC):
    min. 97.0 %
  • Purity (Nonaqueous Titration):
    min. 97.0 %

Specification Test Results

About this item

 :
 :124.1405 g/mol
Boiling Pt:130°C
Melting Pt:38°C
 :GHS07
CAS Number:10167-97-2
Formula:C₆H₈N₂O
MDL Number:MFCD07374873