

Aprepitant ≥98.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:170729-80-3
:
Formula:C₂₃H₂₁F₇N₄O₃
MDL Number:MFCD08277635
:534.43 g/mol
Merck Index:14,00753
:GHS07
:GHS08
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 97.0 %
- Melting point:250.0 to 255.0 deg-C
- Specific rotation [a]20/D:+67.0 to +71.0 deg (C=1,MeOH)
Specification Test Results
Frequently Bought Together